Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009757.4 | NP_033887.1 | 2 | 881 | ||
GenBank mRNA | AF082348.1 | - | 2 | 764 | ||
AJ010259.1 | - | 1 | 108 | |||
AJ132406.1 | - | 2 | 521 | |||
AK028735.1 | - | 2 | 567 | |||
AK135757.1 | - | 2 | 788 | |||
AK135882.1 | - | 2 | 884 | |||
AK136127.1 | - | 2 | 275 | |||
AK136129.1 | - | 2 | 275 | |||
AK139773.1 | - | 2 | 882 | |||
AK139847.1 | - | 2 | 883 | |||
AK139936.1 | - | 2 | 837 | |||
BC055363.1 | - | 2 | 878 | 30051425 |