Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009760.4 | NP_033890.1 | 6 | 853 | ||
GenBank mRNA | AF041054.1 | - | 6 | 823 | ||
AK014223.1 | - | 6 | 854 | |||
AK075943.1 | - | 6 | 854 | |||
AK152610.1 | - | 6 | 849 | |||
AK170865.1 | - | 6 | 817 | |||
BC033278.1 | - | 6 | 761 | |||
BC046603.1 | - | 6 | 843 | 6390074 |