Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smyd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160127.1 | NP_001153599.1 | 10 | 1780 | ||
NM_009762.2 | NP_033892.2 | 9 | 1741 | |||
GenBank mRNA | AK076266.1 | - | 9 | 1706 | ||
AK142153.1 | - | 9 | 1709 | |||
AK142252.1 | - | 10 | 1758 | |||
BC076601.1 | - | 10 | 1758 | 30609878 | ||
U76373.2 | - | 10 | 1752 | |||
U76374.2 | - | 9 | 1713 |