Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacybp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009786.2 | NP_033916.1 | 3, 4 | 568 | ||
GenBank mRNA | AK013924.1 | - | 3, 4 | 499 | ||
AK089043.1 | - | 3, 4 | 422 | |||
AK145013.1 | - | 3, 4 | 535 | |||
AK212050.1 | - | 1, 2 | 32 | |||
AK215280.1 | - | 1, 2 | 32 | |||
BC025948.1 | - | 3, 4 | 453 | 5150537 | ||
U97327.1 | - | 3, 4 | 398 |