Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009787.2 | NP_033917.2 | 4 | 768 | ||
GenBank mRNA | AK028292.1 | - | 4 | 618 | ||
AK146288.1 | - | 4 | 601 | |||
AK148801.1 | - | 4 | 688 | |||
AK150566.1 | - | 4 | 649 | |||
AK161534.1 | - | 4 | 606 | |||
AK168447.1 | - | 4 | 571 | |||
AK169387.1 | - | 4 | 613 | |||
BC066857.1 | - | 3 | 367 | |||
BC141078.1 | - | 4 | 664 | 9055690 | ||
J05186.1 | - | 4 | 651 | |||
Y00884.1 | - | 3 | 474 |