Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009791.4 | NP_033921.3 | 2 | 539 | ||
GenBank mRNA | AF533752.1 | - | 2 | 517 | ||
AK050785.1 | - | 2 | 535 | |||
AK052326.1 | - | 2 | 540 | |||
AK052515.1 | - | 1 | 2038 | |||
AK079744.1 | - | 2 | 505 | |||
AK083710.1 | - | 2 | 506 | |||
AY971958.1 | - | 2 | 380 |