Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009793.3 | NP_033923.2 | 10 | 1213 | ||
GenBank mRNA | AK030219.1 | - | 10 | 1097 | ||
AK043114.1 | - | 10 | 923 | |||
BC070420.1 | - | 10 | 1094 | 5718423 | ||
J03057.1 | - | 2 | 116 | |||
M64266.1 | - | 2 | 164 | |||
X58995.1 | - | 10 | 903 |