Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009794.3 | NP_033924.2 | 10 | 1289 | ||
GenBank mRNA | AF015038.1 | - | 10 | 1235 | ||
AK053832.1 | - | 10 | 2575 | |||
AK078787.1 | - | 6 | 1918 | |||
AK150695.1 | - | 10 | 1263 | |||
AK152094.1 | - | 10 | 1263 | |||
AK152790.1 | - | 10 | 1266 | |||
AK152994.1 | - | 10 | 1266 | |||
AK154855.1 | - | 10 | 1303 | |||
BC054726.1 | - | 10 | 1289 | 5703563 | ||
D38117.1 | - | 10 | 1330 | |||
Y10139.1 | - | 10 | 1171 |