Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083957.1 | NP_001077426.1 | 7 | 918 | ||
NM_009799.4 | NP_033929.2 | 8 | 930 | |||
GenBank mRNA | AK003066.1 | - | 8 | 931 | ||
AK033719.1 | - | 7 | 920 | |||
AK146372.1 | - | 8 | 931 | |||
AK162331.1 | - | 7 | 918 | |||
AK167969.1 | - | 8 | 930 | |||
AK172076.1 | - | 8 | 931 | |||
BC011223.1 | - | 7 | 902 | 4208180 | ||
BC110681.1 | - | 7 | 900 | 4984216 | ||
M32452.1 | - | 7 | 887 |