Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005259.2 | NP_005250.1 | 2 | 695 | ||
GenBank mRNA | AF019627.1 | - | 2 | 562 | ||
AF104922.1 | - | 2 | 695 | |||
AK291524.1 | - | 2 | 666 | |||
BC074757.2 | - | 2 | 609 | 30915216 |