Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009807.2 | NP_033937.2 | 6 | 1033 | ||
GenBank mRNA | AK132826.1 | - | 5 | 1333 | ||
AK163069.1 | - | 3 | 1957 | |||
BC008152.1 | - | 6 | 1033 | 3583883 | ||
L03799.1 | - | 5 | 835 | |||
L28095.1 | - | 6 | 879 |