Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284409.1 | NP_001271338.1 | 6, 7 | 835 | ||
NM_009810.3 | NP_033940.1 | 6, 7 | 830 | |||
GenBank mRNA | AK080651.1 | - | 3, 4 | 345 | ||
AK146226.1 | - | 6, 7 | 837 | |||
BC038825.1 | - | 6, 7 | 719 | 4482368 | ||
D86352.1 | - | 6, 7 | 649 | |||
U19522.1 | - | 6, 7 | 646 | |||
U49929.1 | - | 6, 7 | 672 | |||
U63720.1 | - | 3, 4 | 438 | |||
Y13086.1 | - | 6, 7 | 676 |