Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011710.1 | - | 6, 7 | 657 | ||
AK146363.1 | - | 6, 7 | 667 | |||
AK164321.1 | - | 6, 7 | 633 | |||
AK165998.1 | - | 6, 7 | 644 | |||
AK168842.1 | - | 6, 7 | 665 | |||
BC002022.1 | - | 6, 7 | 653 | 3491126 | ||
Y13087.1 | - | 5, 6 | 621 |