Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011710.1 | - | 7 | 1057 | ||
AK146363.1 | - | 7 | 1067 | |||
AK164321.1 | - | 7 | 1033 | |||
AK165998.1 | - | 7 | 1044 | |||
AK168842.1 | - | 7 | 1065 | |||
BC002022.1 | - | 7 | 1053 | 3491126 | ||
Y13087.1 | - | 6 | 1021 |