Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080126.1 | NP_001073595.1 | 9 | 1912 | ||
NM_001277926.1 | NP_001264855.1 | 10 | 2028 | |||
NM_009812.2 | NP_033942.1 | 9 | 2085 | |||
GenBank mRNA | AF067834.1 | - | 9 | 1965 | ||
AK146758.1 | - | 9 | 2085 | |||
AK168812.1 | - | 9 | 2085 |