Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301153.1 | NP_001288082.1 | 8 | 716 | ||
NM_001301155.1 | NP_001288084.1 | 6 | 447 | |||
NM_001301156.1 | NP_001288085.1 | 5 | 390 | |||
NM_001301157.1 | NP_001288086.1 | 5 | 390 | |||
NM_009817.4 | NP_033947.1 | 7 | 476 | |||
GenBank mRNA | AB026997.1 | - | 8 | 715 | ||
AK028841.1 | - | 7 | 456 | |||
AK028843.1 | - | 7 | 438 | |||
AK029293.1 | - | 6 | 363 | |||
AK036576.1 | - | 5 | 261 | |||
AK083310.1 | - | 5 | 293 | |||
AK083395.1 | - | 5 | 282 | |||
AK089108.1 | - | 5 | 288 | |||
BC010603.1 | - | 5 | 244 | 3710078 | ||
CD550676.1 | - | 8 | 533 | |||
CN529927.1 | - | 6 | 296 | |||
X62519.1 | - | 4 | 160 |