Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301153.1 | NP_001288082.1 | 11, 12 | 1053 | ||
NM_001301155.1 | NP_001288084.1 | 9, 10 | 784 | |||
NM_001301156.1 | NP_001288085.1 | 8, 9 | 727 | |||
NM_001301157.1 | NP_001288086.1 | 8, 9 | 727 | |||
NM_009817.4 | NP_033947.1 | 10, 11 | 813 | |||
GenBank mRNA | AB026997.1 | - | 11, 12 | 1052 | ||
AK028841.1 | - | 10, 11 | 793 | |||
AK028843.1 | - | 10, 11 | 775 | |||
AK029293.1 | - | 9, 10 | 700 | |||
AK036576.1 | - | 8, 9 | 598 | |||
AK083310.1 | - | 8, 9 | 630 | |||
AK083395.1 | - | 8, 9 | 619 | |||
AK089108.1 | - | 8, 9 | 625 | |||
BC010603.1 | - | 8, 9 | 581 | 3710078 | ||
CN529927.1 | - | 9, 10 | 633 |