Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005262.2 | NP_005253.3 | 3 | 678 | ||
GenBank mRNA | AF124603.1 | - | 2 | 386 | ||
AF124604.1 | - | 3 | 524 | |||
AF208342.1 | - | 3 | 366 | |||
AF306863.1 | - | 3 | 608 | |||
AY550027.1 | - | 3 | 368 | |||
BC002429.1 | - | 3 | 614 | 3346679 | ||
BC028348.2 | - | 3 | 605 | 3636052 | ||
U31176.1 | - | 3 | 519 |