Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Runx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111021.2 | NP_001104491.1 | 4 | 576 | ||
NM_001111022.2 | NP_001104492.1 | 4 | 576 | |||
NM_001111023.2 | NP_001104493.1 | 2 | 1911 | |||
NM_009821.3 | NP_033951.2 | 2 | 1911 | |||
GenBank mRNA | AK051758.1 | - | 2 | 1449 | ||
AK137493.1 | - | 4 | 547 | |||
AK145098.1 | - | 4 | 569 | |||
AK155262.1 | - | 3 | 1737 | |||
BC069929.1 | - | 4 | 513 | 4008335 | ||
CB056658.1 | - | 2 | 316 | |||
D13802.1 | - | 2 | 589 | |||
D26532.1 | - | 2 | 1908 | |||
W29200.1 | - | 3 | 340 |