Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Runx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111021.2 | NP_001104491.1 | 8 | 1779 | ||
NM_001111022.2 | NP_001104492.1 | 7 | 1587 | |||
NM_001111023.2 | NP_001104493.1 | 6 | 3114 | |||
NM_009821.3 | NP_033951.2 | 5 | 2922 | |||
GenBank mRNA | AK051758.1 | - | 5 | 2460 | ||
AK137493.1 | - | 8 | 1749 | |||
AK153274.1 | - | 1 | 165 | |||
AK165307.1 | - | 8 | 1774 | |||
BC069929.1 | - | 7 | 1523 | 4008335 | ||
D26532.1 | - | 5 | 2919 |