Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cckar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009827.2 | NP_033957.1 | 2 | 546 | ||
GenBank mRNA | AK004730.2 | - | 2 | 404 | ||
AK143895.1 | - | 2 | 394 | |||
AK146738.1 | - | 2 | 547 | |||
AK147052.1 | - | 2 | 547 | |||
AK164301.1 | - | 138 | ||||
BC020534.1 | - | 2 | 366 | 4236240 |
Set Membership: |
G-protein coupled receptor |