Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cckar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009827.2 | NP_033957.1 | 5 | 1158 | ||
GenBank mRNA | AK004730.2 | - | 5 | 1016 | ||
AK143895.1 | - | 5 | 1006 | |||
AK146738.1 | - | 5 | 1158 | |||
AK147052.1 | - | 5 | 1158 | |||
AK164301.1 | - | 750 |
Set Membership: |
G-protein coupled receptor |