Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005275.3 | NP_005266.2 | 11 | 2963 | ||
XM_005249015.3 | XP_005249072.1 | 12 | 2426 | |||
GenBank mRNA | AK026391.1 | - | 11 | 1625 | ||
AK293596.1 | - | 5 | 916 | |||
AK298042.1 | - | 11 | 1802 | |||
AK301738.1 | - | 10 | 1358 | |||
AK302480.1 | - | 10 | 1691 | |||
AK316391.1 | - | 11 | 1564 | |||
BC013959.2 | - | 11 | 1612 | 2961417 | ||
BC018366.1 | - | 11 | 2543 | 3924808 | ||
BC063657.1 | - | 8 | 1195 | |||
BT006648.1 | - | 11 | 1572 | |||
L25665.1 | - | 11 | 1522 | |||
X66436.1 | - | 2 | 1844 |