Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009833.1 | NP_033963.1 | 3 | 282 | ||
GenBank mRNA | AF087662.1 | - | 3 | 282 | ||
AF095640.1 | - | 3 | 282 | |||
AF109179.1 | - | 3 | 282 | |||
AF113951.1 | - | 3 | 282 | |||
AK083037.1 | - | 3 | 326 | |||
AK133168.1 | - | 982 | ||||
BC131685.1 | - | 3 | 282 | 40111509 | ||
BC131686.1 | - | 3 | 282 |