Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190333.1 | NP_001177262.1 | 3 | 715 | ||
NM_001190334.1 | NP_001177263.1 | 4 | 587 | |||
NM_001190335.1 | NP_001177264.1 | 3 | 496 | |||
NM_001190336.1 | NP_001177265.1 | 3 | 472 | |||
NM_001190337.1 | NP_001177266.1 | 2 | 381 | |||
NM_001190338.1 | NP_001177267.1 | 2 | 468 | |||
NM_009835.4 | NP_033965.1 | 6 | 917 | |||
GenBank mRNA | AB009369.1 | - | 2 | 471 | ||
AB016031.1 | - | 3 | 546 | |||
AK089712.1 | - | 6 | 918 | |||
AK154413.1 | - | 5 | 1290 | |||
BC100756.1 | - | 4 | 583 | 40046797 | ||
BC100757.1 | - | 3 | 492 | 40046796 | ||
BC100758.1 | - | 3 | 468 | 40046794 | ||
BC105669.1 | - | 2 | 374 | 40046795 |