Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctla4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281976.1 | NP_001268905.1 | 3 | 761 | ||
NM_009843.4 | NP_033973.2 | 4 | 871 | |||
GenBank mRNA | AK138891.1 | - | 4 | 805 | ||
BC042741.1 | - | 4 | 871 | 4936052 | ||
BC052683.1 | - | 4 | 791 | 30072821 | ||
X05719.1 | - | 4 | 782 |