Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009849.2 | NP_033979.2 | 5 | 736 | ||
GenBank mRNA | AF042811.1 | - | 5 | 660 | ||
AK002553.1 | - | 5 | 708 | |||
AK172281.1 | - | 5 | 684 | |||
AY376711.1 | - | 5 | 669 | |||
BC011241.1 | - | 5 | 668 | 3582550 | ||
U91511.1 | - | 4 | 543 |