Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291058.1 | NP_001277987.1 | 3 | 692 | ||
GenBank mRNA | AK002264.1 | - | 3 | 650 | ||
AK170443.1 | - | 3 | 616 | |||
BC021637.1 | - | 3 | 633 | 3981024 | ||
BY207885.1 | - | 2 | 183 | |||
DQ167574.1 | - | 2 | 490 | |||
X68273.1 | - | 3 | 675 |