Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005313.4 | NP_005304.3 | 2, 3 | 402 | ||
GenBank mRNA | AK075455.1 | - | 3, 4 | 516 | ||
AK075481.1 | - | 3, 4 | 303 | |||
AK293249.1 | - | 2 | 75 | |||
BC014433.1 | - | 2, 3 | 343 | 3542405 | ||
BC036000.1 | - | 2, 3 | 356 | 4712175 | ||
BC071878.1 | - | 2, 3 | 345 | 6380973 | ||
BG826781.1 | - | 2, 3 | 403 | |||
D16234.1 | - | 2, 3 | 321 | |||
D83485.1 | - | 2, 3 | 276 | |||
EU668338.1 | - | 2, 3 | 356 | |||
FJ224330.1 | - | 2, 3 | 345 | |||
U42068.1 | - | 2, 3 | 332 | |||
Z49835.1 | - | 2, 3 | 360 |