Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161623.1 | NP_001155095.1 | 13 | 1168 | ||
NM_009862.2 | NP_033992.2 | 15 | 1545 | |||
GenBank mRNA | AF081536.1 | - | 15 | 1545 | ||
AF081537.1 | - | 14 | 1244 | |||
AF081538.1 | - | 14 | 1244 | |||
AF081539.1 | - | 2 | 129 | |||
AF098068.1 | - | 14 | 1291 | |||
AJ223729.1 | - | 14 | 1265 | |||
AK147013.1 | - | 14 | 1262 | |||
AK167850.1 | - | 13 | 1156 | |||
AK210447.1 | - | 2 | 73 | |||
BC006856.1 | - | 14 | 1260 | |||
BC028635.1 | - | 14 | 1298 | 1262927 |