Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271566.1 | NP_001258495.1 | 2 | 444 | ||
NM_001271567.1 | NP_001258496.1 | 2 | 275 | |||
NM_001271568.1 | NP_001258497.1 | 2 | 275 | |||
NM_009863.3 | NP_033993.2 | 2 | 275 | |||
GenBank mRNA | AB018574.1 | - | 2 | 276 | ||
AB018575.1 | - | 2 | 276 | |||
AB018576.1 | - | 1 | 86 | |||
AB018577.1 | - | 1 | 86 | |||
AJ007661.1 | - | 2 | 227 | |||
AK012986.1 | - | 2 | 258 | |||
AK145921.1 | - | 2 | 201 | |||
AK146009.1 | - | 2 | 446 | |||
AK163921.1 | - | 245 | ||||
BC062149.1 | - | 2 | 205 | |||
BC072666.1 | - | 2 | 216 | 30611704 | ||
BC080702.1 | - | 2 | 228 | 30474846 |