Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271566.1 | NP_001258495.1 | 3 | 496 | ||
NM_001271567.1 | NP_001258496.1 | 3 | 327 | |||
NM_001271568.1 | NP_001258497.1 | 3 | 327 | |||
NM_009863.3 | NP_033993.2 | 3 | 327 | |||
GenBank mRNA | AB018574.1 | - | 3 | 328 | ||
AB018575.1 | - | 3 | 328 | |||
AB018576.1 | - | 2 | 138 | |||
AB018577.1 | - | 2 | 138 | |||
AJ007661.1 | - | 3 | 279 | |||
AK012986.1 | - | 3 | 310 | |||
AK145921.1 | - | 3 | 253 | |||
AK146009.1 | - | 3 | 498 | |||
AK163921.1 | - | 297 | ||||
BC062149.1 | - | 3 | 257 | |||
BC072666.1 | - | 3 | 268 | 30611704 | ||
BC080702.1 | - | 3 | 280 | 30474846 |