Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009874.3 | NP_034004.2 | 2 | 212 | ||
GenBank mRNA | AK039084.1 | - | 2 | 200 | ||
AK145759.1 | - | 2 | 138 | |||
AK168286.1 | - | 2 | 211 | |||
BC004605.1 | - | 2 | 170 | 3585145 | ||
BC068160.1 | - | 2 | 197 | 30358255 | ||
BC141043.1 | - | 2 | 169 | 9055655 | ||
CJ111227.1 | - | 2 | 214 | |||
U11822.1 | - | 2 | 175 | |||
X74145.1 | - | 2 | 194 |