Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009874.3 | NP_034004.2 | 11, 12 | 1110 | ||
GenBank mRNA | AK145759.1 | - | 11, 12 | 1036 | ||
AK168286.1 | - | 11, 12 | 1109 | |||
BC004605.1 | - | 10, 11 | 957 | 3585145 | ||
BC068160.1 | - | 11, 12 | 1095 | 30358255 | ||
BC141043.1 | - | 11, 12 | 1067 | 9055655 | ||
U11822.1 | - | 11, 12 | 1073 | |||
X74145.1 | - | 11, 12 | 1092 |