Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009879.3 | NP_034009.2 | 3 | 294 | ||
GenBank mRNA | AF354757.1 | - | 3 | 294 | ||
AK013424.1 | - | 3 | 288 | |||
AK161375.1 | - | 3 | 283 | |||
BC055721.1 | - | 3 | 251 | 6810406 | ||
Y10495.2 | - | 3 | 294 |