Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009880.3 | NP_034010.3 | 3 | 1548 | ||
GenBank mRNA | BC019986.1 | - | 3 | 1548 | 4224352 |