Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009897.2 | NP_034027.1 | 9 | 1427 | ||
GenBank mRNA | AK005076.1 | - | 9 | 1426 | ||
AK131953.1 | - | 9 | 1431 | |||
BC025976.1 | - | 10 | 1342 | 4216758 | ||
Z13968.1 | - | 9 | 1427 |