Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301374.1 | NP_001288303.1 | 5 | 855 | ||
NM_009898.3 | NP_034028.1 | 5 | 768 | |||
GenBank mRNA | AF047388.1 | - | 4 | 602 | ||
AF143955.1 | - | 5 | 739 | |||
AF495468.1 | - | 5 | 706 | |||
AK040885.1 | - | 5 | 712 | |||
AK075799.1 | - | 5 | 703 | |||
AK146290.1 | - | 5 | 706 | |||
AK150962.1 | - | 5 | 710 | |||
AK151755.1 | - | 5 | 710 | |||
AK155527.1 | - | 5 | 711 | |||
AK155860.1 | - | 5 | 706 | |||
AK165296.1 | - | 5 | 710 | |||
AK172447.1 | - | 5 | 706 | |||
BC002136.1 | - | 5 | 696 | 3484185 | ||
BC053398.1 | - | 5 | 686 | 30076939 | ||
U89399.1 | - | 1 | 138 |