Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009906.5 | NP_034036.1 | 13 | 2134 | ||
GenBank mRNA | AF111172.1 | - | 13 | 2142 | ||
AK002418.1 | - | 13 | 2153 | |||
AK048279.1 | - | 13 | 2135 | |||
AK083832.1 | - | 12 | 3399 | |||
AK146759.1 | - | 13 | 2128 | |||
AK155021.1 | - | 13 | 2153 | |||
AK165669.1 | - | 13 | 2160 | |||
AK166648.1 | - | 11 | 1927 | |||
AK169924.1 | - | 13 | 2141 | |||
AK170781.1 | - | 13 | 2130 | |||
BC024820.1 | - | 13 | 2136 | 5360558 |