Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009908.2 | NP_034038.2 | 6, 7 | 1039 | ||
GenBank mRNA | AJ006215.1 | - | 6, 7 | 952 | ||
AK087150.1 | - | 3, 4 | 341 | |||
AK155730.1 | - | 6, 7 | 992 | |||
BC004606.1 | - | 1, 2 | 156 | |||
BC031500.1 | - | 6, 7 | 968 | 2647633 | ||
BC063776.1 | - | 6, 7 | 968 | 6398450 |