Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009919.2 | NP_034049.2 | 2 | 201 | ||
GenBank mRNA | AF022811.1 | - | 2 | 192 | ||
AK002293.1 | - | 2 | 169 | |||
AK011200.1 | - | 2 | 154 | |||
AK152432.1 | - | 2 | 152 | |||
AK154479.1 | - | 2 | 152 | |||
AK168154.1 | - | 2 | 152 | |||
BC034868.1 | - | 2 | 161 | 2938052 | ||
DV653213.1 | - | 2 | 255 |