Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109991.1 | NP_001103461.1 | 36 | 3772 | ||
NM_009929.3 | NP_034059.2 | 37 | 3188 | |||
GenBank mRNA | BC043697.1 | - | 36 | 3772 | ||
BC062931.1 | - | 37 | 3155 | |||
BC064817.1 | - | 37 | 3159 | 6332464 | ||
BC066080.1 | - | 37 | 3154 | 30355615 | ||
D17546.1 | - | 35 | 2582 | |||
L22545.1 | - | 35 | 3026 | |||
U03714.1 | - | 7 | 608 |