Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109991.1 | NP_001103461.1 | 39 | 4231 | ||
NM_009929.3 | NP_034059.2 | 40 | 3647 | |||
GenBank mRNA | AF257775.1 | - | 2 | 173 | ||
AK014292.1 | - | 1 | 103 | |||
BC043697.1 | - | 39 | 4231 | |||
BC062931.1 | - | 40 | 3614 | |||
BC064817.1 | - | 40 | 3618 | 6332464 | ||
BC066080.1 | - | 40 | 3613 | 30355615 | ||
D17546.1 | - | 38 | 3041 | |||
L22545.1 | - | 38 | 3485 | |||
U03714.1 | - | 10 | 1067 |