Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009930.2 | NP_034060.2 | 41, 42 | 3250 | ||
GenBank mRNA | AK019448.2 | - | 7, 8 | 615 | ||
AK029212.1 | - | 41, 42 | 3153 | |||
AK041115.1 | - | 41, 42 | 3158 | |||
AK048546.1 | - | 41, 42 | 3158 | |||
AK147542.1 | - | 41, 42 | 3156 | |||
AK157762.1 | - | 41, 42 | 3158 | |||
AK159763.1 | - | 41, 42 | 3159 | |||
AK159920.1 | - | 41, 42 | 3158 | |||
AK159985.1 | - | 41, 42 | 3158 | |||
AK160048.1 | - | 41, 42 | 3158 | |||
AK160107.1 | - | 41, 42 | 3157 | |||
AK160818.1 | - | 41, 42 | 3158 | |||
AK220558.1 | - | 41, 42 | 3173 | |||
BC013626.1 | - | 32, 33 | 2309 | |||
BC028248.1 | - | 32, 33 | 2314 | |||
BC039923.1 | - | 20, 21 | 1549 | |||
BC043089.1 | - | 41, 42 | 3125 | 6418299 | ||
BC052398.1 | - | 41, 42 | 3142 | 5720872 | ||
BC058724.1 | - | 41, 42 | 3159 | 5705762 |