Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009930.2 | NP_034060.2 | 49 | 4151 | ||
GenBank mRNA | AK019448.2 | - | 15 | 1516 | ||
AK029212.1 | - | 49 | 4057 | |||
AK029451.1 | - | 2 | 204 | |||
AK041115.1 | - | 49 | 4059 | |||
AK048546.1 | - | 49 | 4059 | |||
AK079113.1 | - | 4 | 563 | |||
AK147542.1 | - | 49 | 4057 | |||
AK157762.1 | - | 49 | 4059 | |||
AK159763.1 | - | 49 | 4060 | |||
AK159868.1 | - | 5 | 697 | |||
AK159920.1 | - | 49 | 4060 | |||
AK159985.1 | - | 49 | 4060 | |||
AK160048.1 | - | 49 | 4060 | |||
AK160107.1 | - | 49 | 4058 | |||
AK160818.1 | - | 49 | 4062 | |||
AK220558.1 | - | 49 | 4074 | |||
BC013626.1 | - | 40 | 3210 | |||
BC028248.1 | - | 40 | 3215 | |||
BC039923.1 | - | 28 | 2450 | |||
BC043089.1 | - | 49 | 4026 | 6418299 | ||
BC052398.1 | - | 49 | 4043 | 5720872 | ||
BC058724.1 | - | 49 | 4060 | 5705762 |