Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009933.4 | NP_034063.1 | 35 | 3953 | ||
GenBank mRNA | AK027944.1 | - | 1 | 734 | ||
AK029428.1 | - | 1 | 708 | |||
BC002194.1 | - | 6 | 1966 | |||
X66405.1 | - | 35 | 3954 | |||
Z18271.1 | - | 16 | 2492 |