Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009938.4 | NP_034068.3 | 15, 16 | 1829 | ||
GenBank mRNA | AK043479.1 | - | 15, 16 | 1555 | ||
AK183333.1 | - | 2, 3 | 197 | |||
BC082785.1 | - | 11, 12 | 1169 |