Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009948.2 | NP_034078.2 | 5, 6 | 639 | ||
GenBank mRNA | AB010826.1 | - | 4, 5 | 574 | ||
AF017174.2 | - | 5, 6 | 639 | |||
AK029141.1 | - | 5, 6 | 615 | |||
AK146851.1 | - | 5, 6 | 641 | |||
BC018270.1 | - | 5, 6 | 606 | 4920625 | ||
CJ092541.1 | - | 5, 6 | 639 |