Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009950.2 | NP_034080.1 | 3 | 654 | ||
GenBank mRNA | AF017992.1 | - | 2 | 535 | ||
AJ224738.1 | - | 3 | 653 | |||
AJ224739.1 | - | 2 | 390 | |||
AJ224740.1 | - | 3 | 639 | |||
AK136561.1 | - | 3 | 559 | |||
AK163112.1 | - | 3 | 666 | |||
BC005608.1 | - | 2 | 617 | 3498522 |