Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009950.2 | NP_034080.1 | 3 | 1305 | ||
GenBank mRNA | AJ224738.1 | - | 3 | 1304 | ||
AK136561.1 | - | 3 | 1210 | |||
BC005608.1 | - | 2 | 1268 | 3498522 |